Nile red ≥95% (by HPLC), Ultrapure
Catalog # ENZOENZ52551
Supplier: ENZO LIFE SCIENCES
New Product
CAS Number:
Specifications
- Pk:25 mg
- Seq:2315
- Environmentally Preferable:
Specifications
About this item
Synonyms: 9-(Diethylamino)-5H-benzo[a]phenoxazin-5-one
:
:318.38 g/mol
Storage Temperature:Freezer
CAS Number:7385-67-3
Formula:C₂₀H₁₈N₂O₂
MDL Number:MFCD00011639