Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Adapalene
Adapalene ≥98%
Adapalene ≥98%
Adapalene ≥98%
Catalog # ENZOBMLGR1110020
Supplier:
Enzo Life Sciences
CAS Number:
Expand image
Adapalene ≥98%
Catalog # ENZOBMLGR1110020
Supplier:
Enzo Life Sciences
CAS Number:
Specifications
Pk:
20 mg
Seq:
2305
Specifications
About this item
Product Details
:
:
412.53 g/mol
CAS Number:
106685-40-9
Formula:
C₂₈H₂₈O₃
MDL Number:
MFCD03106112
Product Details