Order Entry
Czech Republic
ContactUsLinkComponent
13-cis-Retinoic acid ≥95% (by TLC)
Irritant
Health
13-cis-Retinoic acid ≥95% (by TLC)
Catalog # ENZOBMLGR1020500
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Irritant
Health
13-cis-Retinoic acid ≥95% (by TLC)
Catalog # ENZOBMLGR1020500
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    500 mg
  • Seq:
    2725

Specifications

Specification Test Results

  • Purity:
    ≥95%
  • Appearance:
    Orange solid

Specification Test Results

About this item

 :
 :300.44 g/mol
 :GHS07
 :GHS08
Storage Temperature:Freezer
CAS Number:4759-48-2
Formula:C₂₀H₂₈O₂
MDL Number:MFCD00079542