3-Azaspiro[5.5]undecane hydrochloride ≥97% (by HPLC)
Catalog # ENZOBMLCM1280050
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:50 mg
- Seq:2370
Specifications
About this item
:
:189.725 g/mol
Storage Temperature:Ambient
CAS Number:1125-01-5
:180-44-9
Formula:C10H20ClN
MDL Number:MFCD02093801