N-(3-Ethynylphenyl)-6,7-bis(2-methoxyethoxy)quinazolin-4-amine hydrochloride ≥95% (by HPLC)
Catalog # ENZOENZCHM1570050
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:50 mg
- Seq:2370
Specifications
About this item
:
:429.901 g/mol
CAS Number:183319-69-9
Formula:C₂₂H₂₄ClN₃O₄
MDL Number:MFCD07781272