Permethrin (mixture of cis- and trans-isomeres) (synthetic) ≥96% (by HPLC)
Katalogové číslo ENZOBMLPR1030050
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:50 mg
- Seq:2370
Specifications
About this item
:
:391.3 g/mol
Storage Temperature:Ambient
CAS Number:52645-53-1
Formula:C₂₁H₂₀Cl₂O₃