0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Cytochalasin E
Cytochalasin E ≥98% (by HPLC, TLC)
Cytochalasin E ≥98% (by HPLC, TLC)
ENZOBMLCT1200005
:
Enzo Life Sciences
:
Cytochalasin E ≥98% (by HPLC, TLC)
ENZOBMLCT1200005
:
Enzo Life Sciences
:
Pk:
5 mg
Seq:
2210
:
:
495.57 g/mol
:
Freezer
:
36011-19-5
:
C₂₈H₃₃NO₇