Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Linoleic acid
Linoleic acid ≥99%, colourless oil
Linoleic acid ≥99%, colourless oil
ENZOBMLFA0141000
:
ENZO LIFE SCIENCES
:
Linoleic acid ≥99%, colourless oil
ENZOBMLFA0141000
:
ENZO LIFE SCIENCES
:
Pk:
1 g
Seq:
3047
Purity:
≥99%
:
:
280.45 g/mol
:
Freezer
:
60-33-3
:
C₁₈H₃₂O₂
:
MFCD00064241