ContactUsLinkComponent
Gas Filters, Gas Clean kit for 8890 and 8860
Gas Filters, Gas Clean kit for 8890 and 8860
Catalog # VARI179880
Supplier:  VARIAN
Gas Filters, Gas Clean kit for 8890 and 8860
Catalog # VARI179880
Supplier:  VARIAN
Specifications
  • Description:
    Gas clean kit for 8890 and 8860 (includes carrier gas filter, 1/8" connection unit with mounting bracket, and gas clean sensor)
  • Pk:
    1 KIT