Order Entry
Czech Republic
ContactUsLinkComponent
Telescopic Handle
Telescopic Handle
Catalog # PPSA2016114
Telescopic Handle
Catalog # PPSA2016114

Specifications

  • Colour:
    Grey
  • Description:
    Stainless steel telescopic handle CR ultra, short, 390 g weight
  • Dimensions:
    0,7 m
  • Pk:
    1

Specifications

About this item

Ultra lightweight stainless steel telescopic handle with toggle lever, sleeve and quick-release fastener for Clino®Connect and Clino®Connect Lock.

  • Ultralight and stable
  • Height-adjustable
  • Autoclavable

Areas od application: Technical cleanrooms, life science, pharmaceutical cleanrooms, laboratories