WHEATON® Container, Wide Mouth
Container made of polystyrene with foam PE lined polypropylene screw cap.
- Clear polystyrene offers excellent calrity of contents within
- Ideal for the storage of solid samples and powders
- Good resistance to inorganic chemicals
Frequently Bought Together