ContactUsLinkComponent
Quick Charger US for M410 Label Printer
Quick Charger US for M410 Label Printer
Catalog # BRDY176994
Supplier:  Brady
undefined
Quick Charger US for M410 Label Printer
Catalog # BRDY176994
Supplier:  Brady
Specifications
  • Description:
    Quick charger US for M410 label printer, black, 114,30W×69,85H×101,60D mm
  • Pk:
    1
Specifications
About this item
This quick charger charges the rechargeable NiMH battery in the M410 label printer faster than the standard AC adaptor.
  • With a quick charger on hand, you can be sure your battery is ready to go when you need it most
  • Plugs directly into battery and charges battery from a wall outlet rather than from inside the printer
This quick charger charges the rechargeable NiMH battery in the M410 Label Printer faster than the standard AC adaptor. Package includes the Quick Charger only (printer and battery sold separately). With a quick charger on hand, you can be sure your battery is ready to go when you need it most. Plugs directly into battery and charges battery from a wall outlet rather than from inside the printer.
Ordering information: Order reference M410-QC.
Delivery: Package includes the quick charger only (printer and battery sold separately).