Order Entry
Czech Republic
ContactUsLinkComponent
Potassium chloride 0.1 mol/l (0.1 N) in aqueous solution Reag. Ph. Eur. 1069101
Potassium chloride 0.1 mol/l (0.1 N) in aqueous solution Reag. Ph. Eur. 1069101
Catalog # 87898.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Potassium chloride 0.1 mol/l (0.1 N) in aqueous solution Reag. Ph. Eur. 1069101
Catalog # 87898.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :74.55 g/mol
Storage Temperature:Ambient
EINECS:231-211-8
CAS Number:7447-40-7
Formula:KCl
MDL Number:MFCD00011360