Order Entry
Czech Republic
ContactUsLinkComponent
 
Calcium sulphate saturated solution in water Reag. Ph. Eur. 1015201
Catalog # 87803.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Calcium sulphate saturated solution in water Reag. Ph. Eur. 1015201
Catalog # 87803.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :136.14 g/mol
Storage Temperature:Ambient
EINECS:231-900-3
CAS Number:7778-18-9
Formula:CaO₄S
MDL Number:MFCD00010912