Order Entry
Czech Republic
ContactUsLinkComponent
Barium hydroxide 0.05 M Ba(OH)2 (0.1N) volumetric solution, Fluka™
Barium hydroxide 0.05 M Ba(OH)2 (0.1N) volumetric solution, Fluka™
Catalog # 35050-1L
CAS Number:  
Barium hydroxide 0.05 M Ba(OH)2 (0.1N) volumetric solution, Fluka™
Catalog # 35050-1L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Factor:
    0.999 - 1.001
  • BAM/EMPA reference material:
    effective
  • BAM/EMPA Certificate Number:
    effective
  • Traceable to NIST SRM:
    effective

Specification Test Results

About this item

 :
 :171.34 g/mol
Boiling Pt:100 °C (1.013 hPa)
Density:1.010 g/cm3 (20 °C)
Storage Temperature:Ambient
EINECS:231-791-2
CAS Number:17194-00-2
Formula:H₂BaO₂
MDL Number:MFCD00003443