Organic reference standard, β-Alanine 10 µg/ml in methanol
Catalog # PROONE9203
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:107-95-9
- Description:β-Alanine 10 µg/ml in methanol
- Formula:C₃H₇NO₂
- Description:β-Alanine