Order Entry
Czech Republic
ContactUsLinkComponent
Picric acid 10 g/l in aqueous solution Reag. Ph. Eur. 1065801
Picric acid 10 g/l in aqueous solution Reag. Ph. Eur. 1065801
Catalog # 87897.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Picric acid 10 g/l in aqueous solution Reag. Ph. Eur. 1065801
Catalog # 87897.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :229.11 g/mol
EINECS:201-865-9
CAS Number:88-89-1
Formula:C₆H₃N₃O₇
MDL Number:MFCD00007102