Order Entry
Czech Republic
ContactUsLinkComponent
L-Phenylalanine, Sigma-Aldrich®
L-Phenylalanine, Sigma-Aldrich®
Supplier:  Merck
Certificates
L-Phenylalanine, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    27 g/l (20 °C)
  • Melting Point:
    275 - 283 °C (decomposition)
  • Molar Mass:
    165.19 g/mol
  • Bulk Density:
    580 kg/m3
  • pH:
    5.4 - 6.0 (10 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Frequently Bought Together