(S)-(-)-1,2-Epoxypropane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:16088-62-3
:
Formula:C₃H₆O
MDL Number:MFCD00064312
:58.08 g/mol
Boiling Pt:35 °C (1013 hPa)
Melting Pt:–112 °C
Density:0.8268 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:240-241-0
ADR:3,I
:M-Clarity™ quality level = MQ100
UN:1280
Frequently Bought Together