Order Entry
Czech Republic
ContactUsLinkComponent
Potassium Standard Solution
Potassium Standard Solution
Catalog # WTWA252471
Supplier:  WTW
CAS Number:  
Potassium Standard Solution
Catalog # WTWA252471
Supplier:  WTW
CAS Number:  

Specifications

  • Concentration:
    1000 mg/l as K
  • Pk:
    500 mL
  • Description:
    Potassium standard solution, volume: 500 ml, model: SL COD 1000
  • Seq:
    03

Specifications

About this item

Standard solution for accuracy checks in potassium determinations.