Charcoal activated 1 mg/ml in H₂O, light yellow, liquid, Height: 1 - 2 nm, Ø <5 nm
Catalog # SIAL900712-50ML
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mL
- Seq:2370
Specifications
Specification Test Results
- Appearance (Color):Light yellow to yellow to brown
- Appearance (Form):Liquid
- Concentration:1 mg/ml in H₂O
- Emission maximum:520 - 540 nm
- Full width half max (FWHM):70 - 90 nm
Specification Test Results
About this item
:
:12.01 g/mol
CAS Number:7440-44-0
:64365-11-3
Formula:C
MDL Number:MFCD00133992