Zadání objednávky
Czech Republic
ContactUsLinkComponent
Histamine dihydrochloride, bílý, powder
Irritant
Health
Histamine dihydrochloride, bílý, powder
Catalog # CMPLSB49374.100MG
CAS Number:  
Irritant
Health
Histamine dihydrochloride, bílý, powder
Catalog # CMPLSB49374.100MG
CAS Number:  

Specifications

  • Bal.:
    100 mg
  • Sekvence:
    2440

Specifications

About this item

 :
 :184.07 g/mol
Density:0,63 g/cm³
 :GHS07
 :GHS08
CAS Number:56-92-8
Formula:C₅H₁₁Cl₂N₃
MDL Number:MFCD00012703