

1,6-Diaminohexane, fluid
Catalog # CMPLSB29060.1G
Supplier: CUSTOM MADE CHEMICALS LAB
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:116.21 g/mol
Melting Pt:39...40 °C
Density:0,84 g/cm³
Flash Pt:85 °C
UN:2280
ADR:8,III
:GHS05
:GHS07
CAS Number:124-09-4
Formula:C₆H₁₆N₂
MDL Number:MFCD00008243