BNA Surrogates Mixture
Catalog # EHERA50000527DI
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Concentration:4000 µg/ml in dichloromethane
- Pk:1 mL
- Description:HJ 834-2017 8270 BNA surrogates mixture 527
- Shipping conditions:+20°C
- Storage temperature:−18 °C
Specifications
About this item
Stable isotope labelled product type, mixture format, dichloromethane solvent.