Order Entry
Czech Republic
ContactUsLinkComponent
 
TickBorne Encephalitis Virus NS1 (from HEK293 cells)
  AVIVOPNB00054-100U
 :  
TickBorne Encephalitis Virus NS1 (from HEK293 cells)
  AVIVOPNB00054-100U
 :  

 

  • Pk:
    100 µG
  • Source:
    HEK293 cells
  • Storage conditions:
    Store short-term at 4 °C for up to 1 week. Store long-term at −80 °C. Stable at <−20°C for 1 year. Avoid repeated freeze/thaw cycles.
  • Form:
    Liquid DPBS
  • Protein/peptide name:
    NS1
  • Purity:
    >90%
  • Concentration:
    Varies by lot. See vial for concentration.
  • pH:
    7,4

 

 

TickBorne Encephalitis virus NS1 contains a C-terminal 6x his-tag with protein range 773 to 1128aa.

  • Strain: Neudoerfl
 : Research use only.