Order Entry
Czech Republic
ContactUsLinkComponent
 
Temperature Sensor for CVS, PT1000
  METR6.1110.120
 :  METROHM
Temperature Sensor for CVS, PT1000
  METR6.1110.120
 :  METROHM

 

  • Range:
    −50...+180 °C
  • Description:
    PT1000 temperature sensor with glass shaft and flexible SGJ sleeve, Metrohm plug-in head G
  • Pk:
    1

 

 

PT1000 temperature sensor (class B) for professional CVS instruments.

  • Shaft diameter top: 12 mm
  • Shaft diameter bottom: 5 mm
  • Minimum immersion depth: 20 mm