ContactUsLinkComponent
CPC® NSH Quick Fittings, ³/₈" Hose Barb Valved In-Line Couplings
CPC® NSH Quick Fittings, ³/₈" Hose Barb Valved In-Line Couplings
Supplier:  CPC COLDER
Certificates
CPC® NSH Quick Fittings, ³/₈" Hose Barb Valved In-Line Couplings
Supplier:  CPC COLDER
Specifications
  • Catalog No:
  • CPCCNSHD22006
  • CPCCNSHD17006
  • Max. operating pressure:
  • Vacuum to 120 psi, 8,3 bar
  • Vacuum to 120 psi, 8,3 bar
  • Operating temperature:
  • 0...49 °C
  • 0...49 °C