Order Entry
Czech Republic
ContactUsLinkComponent
Tris(3-hydroxypropyltriazolylmethyl)amine ≥95%
Irritant
Tris(3-hydroxypropyltriazolylmethyl)amine ≥95%
Catalog # CAYM35670-100
Supplier:  Cayman Chemical
CAS Number:  
Irritant
Tris(3-hydroxypropyltriazolylmethyl)amine ≥95%
Catalog # CAYM35670-100
Supplier:  Cayman Chemical
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :434.51 g/mol
 :GHS07
Storage Temperature:Room Temperature
CAS Number:760952-88-3
Formula:C₁₈H₃₀N₁₀O₃
MDL Number:MFCD27665386