Order Entry
Czech Republic
ContactUsLinkComponent
Tissue Genomic DNA Extraction Kits
Tissue Genomic DNA Extraction Kits
Supplier:  BIOPREMIER
Certificates
Tissue Genomic DNA Extraction Kits
Supplier:  BIOPREMIER

Specifications

  • Application:
  • Method / Format:
  • Refine target molecule:
  • Sample type:
  • Target molecule:
  • Environmentally Preferable:

Specifications

Frequently Bought Together