Nitrile Foam Impact Gloves, VHR15
Supplier: Portwest
Certificates
About this item
Innovative TPR design combined with a level F cut liner makes for an impact glove with exceptional dexterity, comfort, and protection.
- Maximum impact protection using TPR pod technology
- Level F cut resistance
- Reinforced thumb crotch and padded palms
- Wrist strap for secure fitting
- Protection against contact heat up to 100 °C
- Nitrile foam coating for excellent grip in wet and dry conditions
- Double-dipped coating for durability
VHR15 Nitrile foam impact gloves are constructed from a composition of TPR, HPPE, polyester, elastane, nitrile foam, and nitrile. The breathable 15 gauge liner ensures outstanding dexterity for the wearer. A double-dipped nitrile foam coating offers excellent grip in wet and dry conditions. A reinforced thumb crotch, as well as exterior and interior padded palms provide increased durability and comfort.
Suitable for use with touchscreen devices. Designed specifically for the oil and gas industry.
Certifications: EN ISO 21420: 2020 Dexterity 5. EN 388: 2016 + A1: 2018 4X42FP. EN 407: 2020 X1XXXX. ANSI/ISEA 105: 2016 CUT Level A7. ANSI/ISEA 105: 2016 ABRASION Level 6. ANSI/ISEA 138: 2019 Level 2.
Specifications
- Catalog No:
- PRTWA755K3RXXL
- PRTWA755K3RXL
- PRTWA755K3RS
- PRTWA755K3RM
- PRTWA755K3RL
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile