Acetic Acid : Triethylamine solution 1:1 2M:2M in aqueous solution, LiChropur™ suitable for HPLC, Supelco®
Catalog # 57619-100ML
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:161.24 g/mol
CAS Number:5204-74-0
Formula:CH₃COOH·N(CH₂CH₃)₃
MDL Number:MFCD00151725