Order Entry
Czech Republic
ContactUsLinkComponent
Sodium dihydrogen phosphate 99.0-101.0% (by titrimetric analysis) suitable for HPLC, Supelco®
Sodium dihydrogen phosphate 99.0-101.0% (by titrimetric analysis) suitable for HPLC, Supelco®
Catalog # 52074-250G
Supplier:  Merck
CAS Number:  
Sodium dihydrogen phosphate 99.0-101.0% (by titrimetric analysis) suitable for HPLC, Supelco®
Catalog # 52074-250G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    250 g
  • Seq:
    3545

Specifications

About this item

 :
 :119.98 g/mol
CAS Number:7558-80-7
Formula:NaH₂PO₄
MDL Number:MFCD00003527