Titanium(IV) chloride 1 mol/l in toluene, AcroSeal®
:7550-45-0
:
:Cl₄Ti
:MFCD00011267
:189.69 g/mol
:0.964 g/cm³
:8 °C
:Ambient
:231-441-9
:3286
:3,II
:GHS02
:GHS05
:GHS06
:GHS08
- Appearance:Clear orange Solution
- Titration Argentometric:0.95 to 1.1 M
Frequently Bought Together