Polypropylene glycol, M.W. ∼4,000, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:25322-69-4
:
Formula:H(OCH₂CH₂CH₂)nOH
MDL Number:MFCD00084446
:1025
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:500-039-8
Frequently Bought Together