Order Entry
Czech Republic
ContactUsLinkComponent
Periodic acid 50%, GPR RECTAPUR®
Periodic acid 50%, GPR RECTAPUR®
Catalog # 294604D
Supplier:  VWR Chemicals
CAS Number:  
undefined
Periodic acid 50%, GPR RECTAPUR®
Catalog # 294604D
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Assay:
    48 - 53 %

Specification Test Results

About this item

 :
 :227.94 g/mol
EINECS:233-937-0
UN:3098
ADR:5.1 (8),II
 :GHS03
 :GHS05
CAS Number:10450-60-9
Formula:H₅IO₆
MDL Number:MFCD00011344