Periodic acid 34% (w/w) in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10450-60-9
:
Formula:H₅IO₆
MDL Number:MFCD00011344
:227.94 g/mol
Merck Index:14,7171
Storage Temperature:Ambient
EINECS:233-937-0
UN:3098
ADR:5.1,II
:GHS03
:GHS05
Related Information
Frequently Bought Together