Order Entry
Czech Republic
ContactUsLinkComponent
Hexane (mixture of isomers) 99+% for HPLC
Flammable
Health
Irritant
Environment
Hexane (mixture of isomers) 99+% for HPLC
Certificates
Flammable
Health
Irritant
Environment
Hexane (mixture of isomers) 99+% for HPLC
Certificates

Specification Test Results

  • Optical Absorbance at 195nm:
    ≤1.00AU
  • Optical Absorbance at 225nm:
    ≤0.050AU
  • Optical Absorbance at 250nm:
    ≤0.010AU
  • Optical Absorbance at 275nm:
    ≤0.005AU
  • Comment:
    Submicron filtered
  • Impurity content (Heptachlorepoxide by GC-ECD responsive residue):
    ≤10 ppt ( single peak)
  • Water Content (Karl Fischer Titration):
    ≤0.01%
  • Optical Absorbance at 300nm:
    ≤0.005AU
  • Residue on Evaporation:
    ≤5 mg/ml
  • Assay (GC):
    ≤60.0% as n-Hexane

Specification Test Results

Related Information

Frequently Bought Together