
Ethanol ≥70% (v/v), irradiated
Supplier: VWR CUSTOM MADE CHEMICALS
Certificates
About this item
CAS Number:64-17-5
:
Formula:C₂H₅OH
MDL Number:MFCD00003568
:46.07 g/mol
ADR:3,III
:Filtered through a 0.2 µm filter.
EINECS:200-578-6
UN:1170
:GHS02
Storage Temperature:Ambient
Related Information
Certificates
Search by Lot Numberspec_VWRS94007.1006 spec_VWRS94007.1016 spec_VWRS94007.5004
Product specification
Frequently Bought Together