Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Xylitol 99+%
Xylitol 99+%
Certificates
Xylitol 99+%

4 Options Available Below

Product Details & Documents
Synonyms:  (2R,3r,4S)-Pentane-1,2,3,4,5-pentaol
  • CAS Number:87-99-0
  • Formula:C₅H₁₂O₅
  • MDL Number:MFCD00064292
  • Molecular Weight:152.15 g/mol
  • Merck Index:13,10140
  • Boiling Pt:215…217 °C
  • Melting Pt:92…96 °C
  • Density:1,52 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:201-788-0

Documents

Certificates
Search by Lot Number
Specifications for Product Family

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    92 to 96 °C
  • Water:
    ≤0.5 %
  • Sulfated ash:
    ≤0.5 %
  • GC:
    ≤2.0 % Other polyols
  • Arsenic (As):
    ≤3 ppm
  • Heavy metals (as Pb):
    ≤10 ppm
  • Reducing sugars:
    ≤0.2 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 1 010,00 Kč
    1 010,00 Kč
    1 * 25 g
       1 010,00 Kč

  • Plastic bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 2 590,00 Kč
    2 590,00 Kč
    1 * 100 g
       2 590,00 Kč

  • Plastic bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 7 010,00 Kč
    7 010,00 Kč
    1 * 500 g
       7 010,00 Kč

  • Plastic bottle
    2,5 kg
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 25 030,00 Kč
    25 030,00 Kč
    1 * 2,5 kg
       25 030,00 Kč

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.