Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
cis-Anethole
cis-Anethole
cis-Anethole
cis-Anethole
PROOMM0164.06
:
LGC Standards PROMOCHEM
:
cis-Anethole
PROOMM0164.06
:
LGC Standards PROMOCHEM
:
Pack type:
Glass bottle for solids
Pk:
100 mg
Seq:
2440
:
:
148.2 g/mol
:
247-181-4
:
25679-28-1
:
C₁₀H₁₂O