Boron trichloride 1 M in hexane, AcroSeal®
:10294-34-5
:
:BCl₃
:MFCD00011313
:117.17 g/mol
:0.738 g/cm³
:–17 °C
:Refrigerator
:233-658-4
:3286
:3,I
:GHS02
:GHS05
:GHS06
:GHS08
:GHS09
- Appearance:Clear colorless to light yellow Solution
- Titration Argentometric:0.9 to 1.1 M
Frequently Bought Together