Order Entry
Czech Republic
ContactUsLinkComponent
Ammonia solution 28% (NH₃ basis) in water ≥99.99% (trace metals basis)
Irritant
Corrosive
Environment
Ammonia solution 28% (NH₃ basis) in water ≥99.99% (trace metals basis)
Certificates
Irritant
Corrosive
Environment
Ammonia solution 28% (NH₃ basis) in water ≥99.99% (trace metals basis)

Specification Test Results

  • Assay (acidimetric):
    28.0 - 30.0 %
  • Metal trace analysis (ICP):
    Max. 0.01 %

Specification Test Results

Related Information

Frequently Bought Together