Order Entry
Czech Republic
ContactUsLinkComponent
 
Irritant
Ammonia solution 0.1 mol/l (0.1 N) Reag. Ph. Eur. 1004704
Catalog # 87767.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Ammonia solution 0.1 mol/l (0.1 N) Reag. Ph. Eur. 1004704
Catalog # 87767.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :35.05 g/mol
EINECS:215-647-6
 :GHS07
Storage Temperature:Ambient
CAS Number:1336-21-6
 :7664-41-7
Formula:NH₃
MDL Number:MFCD00066650