

2-Propanol, Multisolvent® for HPLC
Catalog # SCARAL03212500
Supplier: Scharlau
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
About this item
:
:60.1 g/mol
Merck Index:13,05228
Boiling Pt:82 °C (1013 hPa)
Melting Pt:–89 °C
Density:0.786 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
ADR:3,II
REACH:01-2119457558-25
:GHS02
:GHS07
CAS Number:67-63-0
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674