4-Isopropyl-m-cresol, Sigma-Aldrich®
Catalog # SIAL316431-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:150.22
:M-Clarity™ quality level = MQ100
EINECS:221-761-7
CAS Number:3228-02-2
Formula:C₁₀H₁₄O
MDL Number:MFCD00010704