2-Amino-p-cresol, Sigma-Aldrich®
Catalog # SIAL144908-50G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:123.15 g/mol
Boiling Pt:259 °C (1013 hPa)
Melting Pt:133…136 °C
:M-Clarity™ quality level = MQ100
EINECS:202-457-3
CAS Number:95-84-1
Formula:C₇H₉NO
MDL Number:MFCD00007699