Order Entry
Czech Republic
ContactUsLinkComponent
2,6-Dichloro-p-cresol
2,6-Dichloro-p-cresol
Catalog # EHERC12427100
Supplier:  EHRENSTORFER
CAS Number:  
2,6-Dichloro-p-cresol
Catalog # EHERC12427100
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :177.03 g/mol
EINECS:219-402-4
CAS Number:2432-12-4
Formula:C₇H₆Cl₂O
MDL Number:MFCD00134679