Accessories for Lovibond® Multi Meter, Handheld, SD 40
Supplier: Lovibond
Certificates
Certificates
About this item
Frequently Bought Together
Conductivity standards, KCl, referenced at 25 °C
Supplier: REAGECON
VWR®, Cold Cranking Simulator (CCS) viscosity standards
Supplier: VWR Chemicals
VWR®, Density standards referenced at 25 °C
Supplier: VWR Chemicals
Reagents for chloride determination using Lovibond® MD photometers
Supplier: Lovibond
Nicotine solution, NMR reference standard, Supelco®
Supplier: Merck
2-Naphthylamine, Supelco®
Supplier: Merck
PCB-Mix 1 10 ng/µl 1 * 10 mL
Supplier: EHRENSTORFER
Sigma Pseudo™ corpse scent, drowned victim scent, Supelco®
Supplier: Merck
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals
SULFUR IN 20 CST MINERAL OIL @ 10UG/G 1 * 100 mL
Supplier: LGC Standards PROMOCHEM