



Tri-tert-butylphosphine ≥99%
Supplier: abcr
Certificates
About this item
CAS Number:13716-12-6
:
Formula:C₁₂H₂₇P
MDL Number:MFCD00015006
:202.32 g/mol
Boiling Pt:102…103 °C (13 mmHg)
Melting Pt:30 °C
Density:0.812
Flash Pt:1 °F
EINECS:237-266-4
UN:2845
ADR:4.2,I
:GHS02
:GHS06
:GHS08
:GHS07
Frequently Bought Together