
Dipotassium tetraborate tetrahydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:12045-78-2
:
Formula:B₄K₂O₇·4H₂O
MDL Number:MFCD00149924
:305.49 g/mol
Density:1,95 g/cm³ (20 °C)
EINECS:215-575-5
:GHS08
Frequently Bought Together